siRNA Details
| Gene Symbol: RGD1562101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760641.1 | XP_008758863.1 | 65 | 13442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562101| Gene Symbol: RGD1562101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760641.1 | XP_008758863.1 | 65 | 13442 | ||