siRNA Details
| Gene Symbol: Snrnp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108636.1 | NP_001102106.1 | 6 | 746 | ||
| GenBank mRNA | FQ224589.1 | - | 748 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snrnp27| Gene Symbol: Snrnp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108636.1 | NP_001102106.1 | 6 | 746 | ||
| GenBank mRNA | FQ224589.1 | - | 748 | |||