siRNA Details
| Gene Symbol: Rnf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012195.1 | NP_001012195.1 | 2, 3 | 220 | ||
| GenBank mRNA | BC083614.1 | - | 2, 3 | 220 | 7133774 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf41| Gene Symbol: Rnf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012195.1 | NP_001012195.1 | 2, 3 | 220 | ||
| GenBank mRNA | BC083614.1 | - | 2, 3 | 220 | 7133774 | |