siRNA Details
| Gene Symbol: Ly6e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017467.1 | NP_001017467.1 | 2, 3 | 308 | ||
| XM_006241757.2 | XP_006241819.1 | 3, 4 | 811 | |||
| XM_006241759.2 | XP_006241821.1 | 2, 3 | 448 | |||
| XM_006241760.2 | XP_006241822.1 | 3, 4 | 603 | |||
| XM_006241762.2 | XP_006241824.1 | 2, 3 | 671 | |||
| XM_006241763.2 | XP_006241825.1 | 2, 3 | 668 | |||
| GenBank mRNA | BC079165.1 | - | 2, 3 | 308 | 7129625 | |

Ly6e
Genomic Map