siRNA Details
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766411.1 | XP_008764633.1 | 8 | 1802 | ||
XM_343420.8 | XP_343421.3 | 8 | 1794 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766411.1 | XP_008764633.1 | 8 | 1802 | ||
XM_343420.8 | XP_343421.3 | 8 | 1794 |