siRNA Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766411.1 | XP_008764633.1 | 3 | 1160 | ||
| XM_343420.8 | XP_343421.3 | 3 | 1152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam63b| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766411.1 | XP_008764633.1 | 3 | 1160 | ||
| XM_343420.8 | XP_343421.3 | 3 | 1152 | |||