siRNA Details
| Gene Symbol: Nprl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025744.2 | NP_001020915.2 | 3 | 365 | ||
| GenBank mRNA | BC098932.1 | - | 3 | 344 | ||
| CV077924.1 | - | 3 | 365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nprl2| Gene Symbol: Nprl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025744.2 | NP_001020915.2 | 3 | 365 | ||
| GenBank mRNA | BC098932.1 | - | 3 | 344 | ||
| CV077924.1 | - | 3 | 365 | |||