siRNA Details
| Gene Symbol: LOC100909913 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751998.3 | XP_003752046.1 | 2 | 730 | ||
| Gene Symbol: Ndp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108814.1 | NP_001102284.1 | 683 | |||

LOC100909913
Genomic Map