siRNA Details
| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037546.1 | NP_001032635.1 | 1933 | |||
| XM_008773111.1 | XP_008771333.1 | 2537 | ||||
| GenBank mRNA | BC110051.1 | - | 1933 | 7459258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgd1| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037546.1 | NP_001032635.1 | 1933 | |||
| XM_008773111.1 | XP_008771333.1 | 2537 | ||||
| GenBank mRNA | BC110051.1 | - | 1933 | 7459258 | ||