siRNA Details
| Gene Symbol: RGD1561958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108819.1 | NP_001102289.1 | 2 | 164 | ||
| XM_006257192.2 | XP_006257254.1 | 2 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561958| Gene Symbol: RGD1561958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108819.1 | NP_001102289.1 | 2 | 164 | ||
| XM_006257192.2 | XP_006257254.1 | 2 | 142 | |||