siRNA Details
| Gene Symbol: Olr1481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000527.1 | NP_001000527.1 | 1 | 667 | ||
| GenBank mRNA | M64379.1 | - | 1 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1481| Gene Symbol: Olr1481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000527.1 | NP_001000527.1 | 1 | 667 | ||
| GenBank mRNA | M64379.1 | - | 1 | 91 | ||