siRNA Details
| Gene Symbol: Gtpbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135840.1 | NP_001129312.1 | 4 | 568 | ||
| Gene Symbol: LOC100912590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751210.3 | XP_003751258.2 | 648 | |||

Gtpbp6
Genomic Map