siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014231.1 | NP_001014253.1 | 10 | 1488 | ||
| XM_006250297.2 | XP_006250359.1 | 10 | 1495 | |||
| XM_006250298.2 | XP_006250360.1 | 10 | 2115 | |||
| XM_006250299.2 | XP_006250361.1 | 10 | 1511 | |||
| XM_006250300.2 | XP_006250362.1 | 10 | 1511 | |||
| XM_008769746.1 | XP_008767968.1 | 10 | 1511 | |||
| GenBank mRNA | BC085957.1 | - | 10 | 1488 | 7134976 | |

Dcaf8
Genomic Map