siRNA Details
| Gene Symbol: Nkx2-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127653.1 | NP_001121125.1 | 2 | 433 | ||
| GenBank mRNA | BC160896.1 | - | 2 | 433 | 7453070 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx2-6| Gene Symbol: Nkx2-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127653.1 | NP_001121125.1 | 2 | 433 | ||
| GenBank mRNA | BC160896.1 | - | 2 | 433 | 7453070 | |