siRNA Details
| Gene Symbol: Pdss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014249.1 | NP_001014271.1 | 6 | 1161 | ||
| XM_008773004.1 | XP_008771226.1 | 6 | 1233 | |||
| GenBank mRNA | BC085898.1 | - | 6 | 1161 | 7193369 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdss2| Gene Symbol: Pdss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014249.1 | NP_001014271.1 | 6 | 1161 | ||
| XM_008773004.1 | XP_008771226.1 | 6 | 1233 | |||
| GenBank mRNA | BC085898.1 | - | 6 | 1161 | 7193369 | |