siRNA Details
| Gene Symbol: Rnf224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233651.2 | XP_006233713.1 | 1 | 2137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf224| Gene Symbol: Rnf224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233651.2 | XP_006233713.1 | 1 | 2137 | ||