siRNA Details
| Gene Symbol: Mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108970.1 | NP_001102440.1 | 2 | 817 | ||
| XM_006238001.2 | XP_006238063.1 | 2 | 830 | |||
| XM_008763630.1 | XP_008761852.1 | 2 | 1508 | |||
| XM_008763631.1 | XP_008761853.1 | 4 | 1442 | |||
| XM_008763632.1 | XP_008761854.1 | 3 | 1336 | |||
| XM_008763633.1 | XP_008761855.1 | 2 | 1089 | |||
| XM_008763634.1 | XP_008761856.1 | 2 | 838 | |||
| XM_008763635.1 | XP_008761857.1 | 4 | 1861 | |||
| XM_008763636.1 | XP_008761858.1 | 2 | 1506 | |||
| GenBank mRNA | FQ220583.1 | - | 1 | 308 | ||
| FQ222144.1 | - | 2 | 775 | |||

Mob3b
Genomic Map