siRNA Details
| Gene Symbol: Lsm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108976.1 | NP_001102446.1 | 2 | 808 | ||
| XM_006238895.2 | XP_006238957.1 | 2 | 833 | |||
| XM_006238896.2 | XP_006238958.1 | 2 | 1641 | |||
| XM_006238897.2 | XP_006238959.1 | 2 | 826 | |||
| GenBank mRNA | BC166902.1 | - | 2 | 768 | 6922293 | |

Lsm10
Genomic Map