siRNA Details
| Gene Symbol: Olr987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000590.1 | NP_001000590.1 | 1 | 151 | ||
| Gene Symbol: Olr898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000058.1 | NP_001000058.1 | 1 | 151 | ||

Olr987
Genomic Map