siRNA Details
| Gene Symbol: Pphln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108992.1 | NP_001102462.1 | 9 | 1163 | ||
| XM_006242230.1 | XP_006242292.1 | 9 | 1160 | |||
| XM_006242231.1 | XP_006242293.1 | 8 | 1103 | |||
| XM_006242233.1 | XP_006242295.1 | 8 | 956 | |||
| XM_006242234.1 | XP_006242296.1 | 7 | 899 | |||
| XM_006242235.1 | XP_006242297.1 | 8 | 743 | |||
| XM_006242236.2 | XP_006242298.1 | 9 | 927 | |||
| GenBank mRNA | AY724510.1 | - | 7 | 839 | ||
| BC166484.1 | - | 7 | 692 | |||
| FQ211850.1 | - | 3 | 247 | |||

Pphln1
Genomic Map