siRNA Details
| Gene Symbol: Alg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109000.1 | NP_001102470.1 | 9 | 1025 | ||
| XM_008766322.1 | XP_008764544.1 | 9 | 1603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alg9| Gene Symbol: Alg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109000.1 | NP_001102470.1 | 9 | 1025 | ||
| XM_008766322.1 | XP_008764544.1 | 9 | 1603 | |||