siRNA Details
Gene Symbol: Sowahd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173373.1 | NP_001166844.1 | 1 | 991 | ||
GenBank mRNA | BC162046.1 | - | 1 | 991 | 7107377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sowahd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173373.1 | NP_001166844.1 | 1 | 991 | ||
GenBank mRNA | BC162046.1 | - | 1 | 991 | 7107377 |