siRNA Details
| Gene Symbol: Taf5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107442.1 | NP_001100912.1 | 5 | 1505 | ||
| XM_006255812.2 | XP_006255874.1 | 4 | 2573 | |||
| XM_008772614.1 | XP_008770836.1 | 4 | 1393 | |||
| XM_008772615.1 | XP_008770837.1 | 4 | 1569 | |||
| GenBank mRNA | BC160901.1 | - | 4 | 1407 | 7454090 | |

Taf5l
Genomic Map