siRNA Details
| Gene Symbol: RASSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164746.1 | NP_001158218.1 | 3 | 1432 | ||
| NM_001164747.1 | NP_001158219.1 | 4 | 1177 | |||
| NM_001164748.1 | NP_001158220.1 | 3 | 1111 | |||
| NM_007211.4 | NP_009142.2 | 4 | 1205 | |||
| XM_006719027.2 | XP_006719090.1 | 3 | 979 | |||
| XM_006719028.1 | XP_006719091.1 | 3 | 1383 | |||
| XM_017018746.1 | XP_016874235.1 | 3 | 1019 | |||
| GenBank mRNA | AB093206.1 | - | 3 | 1084 | ||
| AK290055.1 | - | 4 | 1177 | |||
| AK292333.1 | - | 3 | 1426 | |||
| AY665468.1 | - | 3 | 1034 | |||
| BC030021.1 | - | 4 | 1207 | 5266519 | ||

RASSF8
Genomic Map