siRNA Details
| Gene Symbol: Olr582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000662.1 | NP_001000662.1 | 1 | 466 | ||
| XM_006234491.1 | XP_006234553.1 | 1 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr582| Gene Symbol: Olr582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000662.1 | NP_001000662.1 | 1 | 466 | ||
| XM_006234491.1 | XP_006234553.1 | 1 | 490 | |||