siRNA Details
| Gene Symbol: Olr1642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000832.1 | NP_001000832.1 | 1 | 757 | ||
| Gene Symbol: Olr1641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000100.1 | NP_001000100.1 | 1 | 757 | ||

Olr1642
Genomic Map