siRNA Details
| Gene Symbol: Olr1305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000959.1 | NP_001000959.1 | 1 | 115 | ||
| Gene Symbol: Olr1304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000464.1 | NP_001000464.1 | 1 | 115 | ||

Olr1305
Genomic Map