siRNA Details
| Gene Symbol: Olr629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001058.1 | NP_001001058.1 | 1 | 657 | ||
| Gene Symbol: Olr691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000630.1 | NP_001000630.1 | 1 | 657 | ||

Olr629
Genomic Map