siRNA Details
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213561.3 | NP_998726.2 | 703 | |||
| GenBank mRNA | BC093374.1 | - | 630 | 7388599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcf19| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213561.3 | NP_998726.2 | 703 | |||
| GenBank mRNA | BC093374.1 | - | 630 | 7388599 | ||