siRNA Details
| Gene Symbol: Nanog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100781.1 | NP_001094251.1 | 2 | 553 | ||
| XM_006237310.2 | XP_006237372.1 | 2 | 350 | |||
| GenBank mRNA | AB162852.1 | - | 1 | 157 | ||
| AB275459.1 | - | 2 | 350 | |||

Nanog
Genomic Map