siRNA Details
| Gene Symbol: Scgb2b24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577783.2 | XP_577783.2 | 2, 3 | 236 | ||
| GenBank mRNA | GU269242.1 | - | 2, 3 | 236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb2b24| Gene Symbol: Scgb2b24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577783.2 | XP_577783.2 | 2, 3 | 236 | ||
| GenBank mRNA | GU269242.1 | - | 2, 3 | 236 | ||