siRNA Details
| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109099.1 | NP_001102569.1 | 4 | 996 | ||
| XM_006251813.2 | XP_006251875.1 | 5 | 1783 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naa30| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109099.1 | NP_001102569.1 | 4 | 996 | ||
| XM_006251813.2 | XP_006251875.1 | 5 | 1783 | |||