siRNA Details
| Gene Symbol: RGD1565222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109101.1 | NP_001102571.1 | 6 | 1526 | ||
| XM_006252004.2 | XP_006252066.1 | 6 | 1824 | |||
| XM_006252005.2 | XP_006252067.1 | 6 | 1482 | |||
| XM_006252006.2 | XP_006252068.1 | 1442 | ||||

RGD1565222
Genomic Map