siRNA Details
Gene Symbol: RGD1559804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113787.1 | NP_001107259.1 | 3 | 549 | ||
GenBank mRNA | BC158776.1 | - | 3 | 549 | 7133601 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1559804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113787.1 | NP_001107259.1 | 3 | 549 | ||
GenBank mRNA | BC158776.1 | - | 3 | 549 | 7133601 |