siRNA Details
| Gene Symbol: RatNP-3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079898.2 | NP_001073367.2 | 3 | 326 | ||
| GenBank mRNA | U16683.1 | - | 3 | 315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RatNP-3b| Gene Symbol: RatNP-3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079898.2 | NP_001073367.2 | 3 | 326 | ||
| GenBank mRNA | U16683.1 | - | 3 | 315 | ||