siRNA Details
| Gene Symbol: Aaed1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109114.1 | NP_001102584.1 | 3, 4 | 371 | ||
| XM_006253495.2 | XP_006253557.1 | 3, 4 | 371 | |||
| XM_006253497.2 | XP_006253559.1 | 3, 4 | 375 | |||
| XM_008771445.1 | XP_008769667.1 | 3, 4 | 380 | |||

Aaed1
Genomic Map