siRNA Details
| Gene Symbol: Nol7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253849.2 | XP_006253911.1 | 3, 4 | 517 | ||
| GenBank mRNA | FQ210031.1 | - | 3, 4 | 389 | ||
| FQ227425.1 | - | 3, 4 | 385 | |||
| FQ229485.1 | - | 3, 4 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nol7| Gene Symbol: Nol7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253849.2 | XP_006253911.1 | 3, 4 | 517 | ||
| GenBank mRNA | FQ210031.1 | - | 3, 4 | 389 | ||
| FQ227425.1 | - | 3, 4 | 385 | |||
| FQ229485.1 | - | 3, 4 | 399 | |||