siRNA Details
| Gene Symbol: Ccdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574081.6 | XP_574081.4 | 3 | 877 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc3| Gene Symbol: Ccdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574081.6 | XP_574081.4 | 3 | 877 | ||