siRNA Details
| Gene Symbol: RGD1559536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195484.1 | NP_001182413.1 | 2, 3 | 154 | ||
| GenBank mRNA | CB779624.1 | - | 2, 3 | 155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559536| Gene Symbol: RGD1559536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195484.1 | NP_001182413.1 | 2, 3 | 154 | ||
| GenBank mRNA | CB779624.1 | - | 2, 3 | 155 | ||