siRNA Details
| Gene Symbol: RGD1562987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173472.1 | NP_001166943.1 | 2, 3 | 292 | ||
| XM_008772163.1 | XP_008770385.1 | 2, 3 | 270 | |||
| GenBank mRNA | BC162045.1 | - | 2, 3 | 281 | 7107313 | |
| CB568734.1 | - | 2, 3 | 293 | |||
| FQ217314.1 | - | 2, 3 | 290 | |||
| FQ224078.1 | - | 2, 3 | 280 | |||
| FQ228113.1 | - | 2, 3 | 268 | |||
| FQ229472.1 | - | 2, 3 | 250 | |||

RGD1562987
Genomic Map