siRNA Details
| Gene Symbol: RGD1562987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173472.1 | NP_001166943.1 | 3 | 523 | ||
| XM_008772163.1 | XP_008770385.1 | 3 | 501 | |||
| GenBank mRNA | BC162045.1 | - | 3 | 512 | 7107313 | |
| CB568734.1 | - | 3 | 524 | |||
| FQ217314.1 | - | 3 | 521 | |||
| FQ224078.1 | - | 3 | 511 | |||
| FQ228113.1 | - | 3 | 499 | |||
| FQ229472.1 | - | 3 | 481 | |||

RGD1562987
Genomic Map