siRNA Details
| Gene Symbol: Pacrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077677.1 | NP_001071145.1 | 5 | 902 | ||
| GenBank mRNA | BC127534.1 | - | 5 | 902 | 7133203 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pacrg| Gene Symbol: Pacrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077677.1 | NP_001071145.1 | 5 | 902 | ||
| GenBank mRNA | BC127534.1 | - | 5 | 902 | 7133203 | |