siRNA Details
| Gene Symbol: Kctd21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109151.1 | NP_001102621.1 | 1 | 550 | ||
| XM_006229781.2 | XP_006229843.1 | 2 | 621 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd21| Gene Symbol: Kctd21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109151.1 | NP_001102621.1 | 1 | 550 | ||
| XM_006229781.2 | XP_006229843.1 | 2 | 621 | |||