siRNA Details
| Gene Symbol: Cdca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760176.1 | XP_008758398.1 | 6 | 1030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdca5| Gene Symbol: Cdca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760176.1 | XP_008758398.1 | 6 | 1030 | ||