siRNA Details
| Gene Symbol: Calhm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191956.1 | NP_001178885.1 | 2 | 486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calhm3| Gene Symbol: Calhm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191956.1 | NP_001178885.1 | 2 | 486 | ||