siRNA Details
| Gene Symbol: LOC499541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047942.1 | NP_001041407.1 | 1, 2 | 94 | ||
| XM_008760791.1 | XP_008759013.1 | 4, 5 | 1022 | |||
| GenBank mRNA | AY383700.1 | - | 1, 2 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499541| Gene Symbol: LOC499541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047942.1 | NP_001041407.1 | 1, 2 | 94 | ||
| XM_008760791.1 | XP_008759013.1 | 4, 5 | 1022 | |||
| GenBank mRNA | AY383700.1 | - | 1, 2 | 94 | ||