siRNA Details
Gene Symbol: LOC499542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047943.1 | NP_001041408.1 | 3 | 294 | ||
GenBank mRNA | AY539929.1 | - | 3 | 294 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC499542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047943.1 | NP_001041408.1 | 3 | 294 | ||
GenBank mRNA | AY539929.1 | - | 3 | 294 |