siRNA Details
| Gene Symbol: LOC499565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047946.1 | NP_001041411.1 | 3 | 293 | ||
| GenBank mRNA | AY383693.1 | - | 3 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC499565| Gene Symbol: LOC499565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047946.1 | NP_001041411.1 | 3 | 293 | ||
| GenBank mRNA | AY383693.1 | - | 3 | 293 | ||