siRNA Details
| Gene Symbol: Fabp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134614.1 | NP_001128086.1 | 3 | 464 | ||
| XM_006232145.2 | XP_006232207.1 | 3 | 1068 | |||
| XM_006232146.2 | XP_006232208.1 | 3 | 597 | |||
| XM_006232147.2 | XP_006232209.1 | 2 | 265 | |||
| XM_006232148.2 | XP_006232210.1 | 2 | 265 | |||
| GenBank mRNA | EU733648.1 | - | 3 | 365 | ||

Fabp12
Genomic Map