siRNA Details
| Gene Symbol: Klrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029908.1 | NP_001025079.1 | 1 | 102 | ||
| XM_008763323.1 | XP_008761545.1 | 1 | 472 | |||
| XM_008763324.1 | XP_008761546.1 | 1 | 472 | |||
| GenBank mRNA | AY197774.1 | - | 1 | 102 | ||
| Gene Symbol: Klrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019261.1 | NP_062134.1 | 1 | 129 | ||
| GenBank mRNA | AF021349.1 | - | 1 | 129 | ||

Klrc3
Genomic Map